Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005550960.2 | XP_005551017.1 | 1 - 2 | 136 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |