Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005561750.2 | XP_005561807.1 | 1 - 2 | 416 | 93 | |
XM_015453990.1 | XP_015309476.1 | 1 - 2 | 443 | 93 | ||
XM_015453991.1 | XP_015309477.1 | 1 - 2 | 430 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |