Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP90AA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283939.1 | NP_001270868.1 | 2 - 3 | 223 | 128 | |
GenBank mRNA | AB169853.1 | - | 2 - 3 | 223 | 128 | |
AB170124.1 | - | 2 - 3 | 223 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |