Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM24B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566658.1 | XP_005566715.1 | 2 - 3 | 310 | 133 | |
XM_015456819.1 | XP_015312305.1 | 3 - 4 | 353 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |