Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591258.2 | XP_005591315.1 | 1 - 2 | 215 | 171 | |
GenBank mRNA | AB169169.1 | - | 1 - 2 | 202 | 171 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |