Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583640.2 | XP_005583697.1 | 2 - 3 | 660 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |