Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568529.2 | XP_005568586.1 | 3 - 4 | 1061 | 50 | |
XM_005568530.2 | XP_005568587.1 | 3 - 4 | 1058 | 50 | ||
GenBank mRNA | AB171090.1 | - | 3 - 4 | 359 | 50 | |
AY650313.1 | - | 1 - 2 | 123 | 50 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |