Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284976.1 | NP_001271905.1 | 6 - 7 | 635 | 57 | |
XM_015434151.1 | XP_015289637.1 | 1 - 2 | 524 | 57 | ||
XM_015434152.1 | XP_015289638.1 | 6 - 7 | 1869 | 57 | ||
XM_015434153.1 | XP_015289639.1 | 6 - 7 | 2149 | 57 | ||
XM_015434154.1 | XP_015289640.1 | 6 - 7 | 1574 | 57 | ||
XM_015434155.1 | XP_015289641.1 | 5 - 6 | 1143 | 57 | ||
XM_015434156.1 | XP_015289642.1 | 5 - 6 | 1038 | 57 | ||
XM_015434157.1 | XP_015289643.1 | 1 - 2 | 524 | 57 | ||
XM_015434158.1 | XP_015289644.1 | 6 - 7 | 1862 | 57 | ||
XM_015434161.1 | XP_015289647.1 | 5 - 6 | 1797 | 57 | ||
XM_015434162.1 | XP_015289648.1 | 1 - 2 | 524 | 57 | ||
GenBank mRNA | AB169413.1 | - | 6 - 7 | 635 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |