Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284989.1 | NP_001271918.1 | 3 - 4 | 460 | 90 | |
XM_005584713.2 | XP_005584770.1 | 2 - 3 | 387 | 90 | ||
XM_015437842.1 | XP_015293328.1 | 3 - 4 | 540 | 90 | ||
GenBank mRNA | AB169157.1 | - | 3 - 4 | 460 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |