Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7H14orf28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015453738.1 | XP_015309224.1 | 4 - 5 | 771 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |