Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015434264.1 | XP_015289750.1 | 6 - 7 | 1074 | 62 | |
XM_015434265.1 | XP_015289751.1 | 7 - 8 | 1217 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |