Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005554304.2 | XP_005554361.1 | 4 - 5 | 1829 | 106 | |
XM_015450034.1 | XP_015305520.1 | 4 - 5 | 2315 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |