Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM20L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005561363.2 | XP_005561420.1 | 1 - 2 | 755 | 69 | |
XM_005561364.2 | XP_005561421.1 | 1 - 2 | 745 | 69 | ||
XM_005561366.2 | XP_005561423.1 | 1 - 2 | 743 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |