Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319434.1 | NP_001306363.1 | 1 - 2 | 1158 | 59 | |
XM_015442462.1 | XP_015297948.1 | 1 - 2 | 1158 | 59 | ||
GenBank mRNA | AB172418.1 | - | 1 - 2 | 1158 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Probe spans exons |