Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005584174.2 | XP_005584231.1 | 6 - 7 | 1288 | 110 | |
XM_005584175.2 | XP_005584232.1 | 6 - 7 | 1241 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |