Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007877.1 | NP_031903.1 | 4 - 5 | 724 | 63 | |
GenBank mRNA | BC119286.1 | - | 4 - 5 | 745 | 8734035 | 63 |
BC137819.1 | - | 4 - 5 | 745 | 8860838 | 63 | |
X67274.1 | - | 4 - 5 | 724 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |