Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008004.4 | NP_032030.1 | 3 - 4 | 761 | 82 | |
GenBank mRNA | AB009250.1 | - | 3 - 4 | 265 | 82 | |
AK077555.1 | - | 3 - 4 | 763 | 82 | ||
AK082858.1 | - | 3 - 4 | 652 | 82 | ||
BC119033.1 | - | 3 - 4 | 747 | 8733782 | 82 | |
BC144776.1 | - | 3 - 4 | 714 | 9053324 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |