Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048177.2 | NP_001041642.1 | 9 - 10 | 1671 | 117 | |
NM_008413.3 | NP_032439.2 | 10 - 11 | 1754 | 117 | ||
GenBank mRNA | AK087255.1 | - | 9 - 10 | 1638 | 117 | |
BC054807.1 | - | 9 - 10 | 1434 | 6838318 | 117 | |
BC059834.1 | - | 9 - 10 | 1434 | 6838318 | 117 | |
L16956.1 | - | 9 - 10 | 1423 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |