Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008431.2 | NP_032457.1 | 1 - 2 | 168 | 107 | |
GenBank mRNA | AF056492.1 | - | 1 - 2 | 206 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mus musculus Probe spans exons |