Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162415.1 | NP_001155887.1 | 12 - 13 | 1325 | 101 | |
NM_001162416.1 | NP_001155888.1 | 10 - 11 | 1118 | 101 | ||
NM_008825.4 | NP_032851.2 | 11 - 12 | 1219 | 101 | ||
NR_027859.1 | 11 - 12 | 1219 | 101 | |||
GenBank mRNA | AK029659.1 | - | 12 - 13 | 1326 | 101 | |
AK146744.1 | - | 11 - 12 | 1213 | 101 | ||
BC018418.1 | - | 11 - 12 | 1176 | 3979400 | 101 | |
BC051014.1 | - | 10 - 11 | 1132 | 6493944 | 101 | |
EU616666.1 | - | 9 - 10 | 992 | 101 | ||
EU616667.1 | - | 10 - 11 | 1117 | 101 | ||
EU616668.1 | - | 10 - 11 | 1117 | 101 | ||
EU616670.1 | - | 9 - 10 | 983 | 101 | ||
X98847.1 | - | 10 - 11 | 1137 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |