Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkci | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008857.3 | NP_032883.2 | 7 - 8 | 855 | 73 | |
GenBank mRNA | AK144979.1 | - | 7 - 8 | 855 | 73 | |
AK147710.1 | - | 7 - 8 | 827 | 73 | ||
AK167411.1 | - | 7 - 8 | 824 | 73 | ||
AK220517.1 | - | 7 - 8 | 879 | 73 | ||
BC021630.1 | - | 7 - 8 | 726 | 3980628 | 73 | |
D28577.1 | - | 7 - 8 | 650 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |