Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008924.2 | NP_032950.1 | 8 - 9 | 1179 | 118 | |
GenBank mRNA | AF533977.1 | - | 8 - 9 | 1179 | 118 | |
AK132728.1 | - | 8 - 9 | 1118 | 118 | ||
AK132824.1 | - | 8 - 9 | 1110 | 118 | ||
BC075623.1 | - | 8 - 9 | 1150 | 6854057 | 118 | |
BC080276.1 | - | 8 - 9 | 1173 | 30354403 | 118 | |
J02935.1 | - | 8 - 9 | 862 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |