Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008941.3 | NP_032967.1 | 21 - 22 | 2480 | 93 | |
NM_178855.4 | NP_849186.2 | 20 - 21 | 2435 | 93 | ||
GenBank mRNA | AK033582.1 | - | 20 - 21 | 2437 | 93 | |
AK038356.1 | - | 12 - 13 | 1429 | 93 | ||
BC117917.1 | - | 21 - 22 | 2479 | 40097367 | 93 | |
BC117918.1 | - | 21 - 22 | 2479 | 40097370 | 93 | |
U73378.1 | - | 21 - 22 | 2475 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |