Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271566.1 | NP_001258495.1 | 6 - 7 | 918 | 78 | |
NM_001271567.1 | NP_001258496.1 | 6 - 7 | 749 | 78 | ||
NM_001271568.1 | NP_001258497.1 | 6 - 7 | 749 | 78 | ||
NM_009863.3 | NP_033993.2 | 6 - 7 | 749 | 78 | ||
GenBank mRNA | AB018574.1 | - | 6 - 7 | 750 | 78 | |
AB018575.1 | - | 6 - 7 | 750 | 78 | ||
AB018576.1 | - | 5 - 6 | 560 | 78 | ||
AB018577.1 | - | 5 - 6 | 560 | 78 | ||
AJ007661.1 | - | 6 - 7 | 701 | 78 | ||
AK145921.1 | - | 6 - 7 | 675 | 78 | ||
AK146009.1 | - | 6 - 7 | 920 | 78 | ||
BC062149.1 | - | 6 - 7 | 679 | 78 | ||
BC072666.1 | - | 6 - 7 | 690 | 30611704 | 78 | |
BC080702.1 | - | 6 - 7 | 702 | 30474846 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |