Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011348.2 | NP_035478.2 | 3 - 4 | 970 | 86 | |
GenBank mRNA | AF034744.1 | - | 3 - 4 | 705 | 86 | |
AK141295.1 | - | 3 - 4 | 972 | 86 | ||
AK142340.1 | - | 3 - 4 | 797 | 86 | ||
AK159214.1 | - | 3 - 4 | 761 | 86 | ||
BC057956.1 | - | 3 - 4 | 868 | 5357516 | 86 | |
Z80941.1 | - | 3 - 4 | 336 | 86 | ||
Z93947.1 | - | 3 - 4 | 945 | 86 | ||
Z93948.1 | - | 3 - 4 | 945 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |