Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011776.1 | NP_035906.1 | 4 - 5 | 751 | 64 | |
GenBank mRNA | AK131934.1 | - | 4 - 5 | 750 | 64 | |
BC099465.1 | - | 4 - 5 | 736 | 30939089 | 64 | |
BC100745.1 | - | 4 - 5 | 751 | 40045702 | 64 | |
BC103583.1 | - | 4 - 5 | 751 | 40045704 | 64 | |
BC103584.1 | - | 4 - 5 | 751 | 40045703 | 64 | |
BC103585.1 | - | 4 - 5 | 751 | 40045701 | 64 | |
M20026.1 | - | 4 - 5 | 751 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |