Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201470.1 | NP_001188399.1 | 8 - 9 | 1206 | 102 | |
NM_011864.3 | NP_035994.2 | 9 - 10 | 1242 | 102 | ||
GenBank mRNA | AF052453.1 | - | 9 - 10 | 1223 | 102 | |
AF085144.1 | - | 8 - 9 | 1138 | 102 | ||
AK128962.1 | - | 8 - 9 | 1227 | 102 | ||
AK137675.1 | - | 8 - 9 | 1132 | 102 | ||
BC090997.1 | - | 9 - 10 | 1242 | 30121331 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |