Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF037205.1 | - | 2 - 3 | 194 | 83 | |
AF037206.1 | - | 2 - 3 | 194 | 83 | ||
AI882163.1 | - | 2 - 3 | 253 | 83 | ||
AK034135.1 | - | 2 - 3 | 245 | 83 | ||
AK036591.1 | - | 2 - 3 | 239 | 83 | ||
AK050095.1 | - | 2 - 3 | 207 | 83 | ||
AK082296.1 | - | 2 - 3 | 235 | 83 | ||
AK139447.1 | - | 2 - 3 | 200 | 83 | ||
AK140682.1 | - | 2 - 3 | 249 | 83 | ||
AK158046.1 | - | 2 - 3 | 235 | 83 | ||
AK167278.1 | - | 2 - 3 | 199 | 83 | ||
BC058182.1 | - | 2 - 3 | 202 | 5324040 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |