Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013484.2 | NP_038512.2 | 11 - 12 | 1584 | 60 | |
GenBank mRNA | AK146812.1 | - | 11 - 12 | 1585 | 60 | |
BC011086.1 | - | 11 - 12 | 1547 | 3992881 | 60 | |
M57891.1 | - | 11 - 12 | 1531 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |