Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083967.1 | NP_001077436.1 | 16 - 17 | 1725 | 98 | |
NM_013685.2 | NP_038713.1 | 16 - 17 | 2040 | 98 | ||
GenBank mRNA | AK014343.1 | - | 9 - 10 | 1109 | 98 | |
AK028748.1 | - | 16 - 17 | 1651 | 98 | ||
AK081012.1 | - | 16 - 17 | 1677 | 98 | ||
AK132140.1 | - | 9 - 10 | 1066 | 98 | ||
AK133885.1 | - | 17 - 18 | 2452 | 98 | ||
AK151433.1 | - | 9 - 10 | 1011 | 98 | ||
BC014293.1 | - | 9 - 10 | 4014231 | 98 | ||
BC043050.1 | - | 15 - 16 | 1500 | 5708370 | 98 | |
S75870.1 | - | 16 - 17 | 1612 | 98 | ||
U16321.1 | - | 10 - 11 | 1099 | 98 | ||
U16322.1 | - | 17 - 18 | 2012 | 98 | ||
X91753.1 | - | 16 - 17 | 1664 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |