Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016811.2 | NP_058091.2 | 3 - 4 | 475 | 95 | |
GenBank mRNA | AF085219.1 | - | 2 - 3 | 161 | 95 | |
BC006713.1 | - | 3 - 4 | 475 | 3584592 | 95 | |
JN832677.1 | - | 4 - 5 | 507 | 95 | ||
JN832678.1 | - | 3 - 4 | 494 | 95 | ||
JN832679.1 | - | 3 - 4 | 494 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |