Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019583.3 | NP_062529.2 | 3 - 4 | 336 | 88 | |
GenBank mRNA | AF208108.1 | - | 3 - 4 | 249 | 88 | |
AF208109.1 | - | 3 - 4 | 249 | 88 | ||
AK045627.1 | - | 3 - 4 | 337 | 88 | ||
AK050139.1 | - | 3 - 4 | 253 | 88 | ||
BC026546.1 | - | 3 - 4 | 236 | 5042466 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |