Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023580.4 | NP_076069.2 | 15 - 16 | 2568 | 83 | |
GenBank mRNA | AF131197.1 | - | 15 - 16 | 2510 | 83 | |
AK028478.1 | - | 15 - 16 | 2569 | 83 | ||
BC071215.1 | - | 15 - 16 | 2532 | 4196138 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |