Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009484.2 | NP_033510.2 | 23 - 24 | 3354 | 119 | |
GenBank mRNA | AF057367.1 | - | 23 - 24 | 3334 | 119 | |
AK142512.1 | - | 23 - 24 | 3270 | 119 | ||
AK162410.1 | - | 21 - 22 | 2927 | 119 | ||
AK164917.1 | - | 10 - 11 | 1799 | 119 | ||
AK220463.1 | - | 24 - 25 | 3311 | 119 | ||
BC053061.1 | - | 23 - 24 | 3336 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |