Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164177.1 | NP_001157649.1 | 1 - 2 | 159 | 78 | |
NM_016883.4 | NP_058579.2 | 1 - 2 | 159 | 78 | ||
GenBank mRNA | AB022022.1 | - | 1 - 2 | 115 | 78 | |
AK009068.1 | - | 1 - 2 | 138 | 78 | ||
AK018233.1 | - | 1 - 2 | 137 | 78 | ||
AK136400.1 | - | 1 - 2 | 138 | 78 | ||
BC026931.1 | - | 1 - 2 | 126 | 3987112 | 78 | |
BC056196.1 | - | 1 - 2 | 144 | 30019985 | 78 | |
BY057263.1 | - | 1 - 2 | 159 | 78 | ||
CD564300.1 | - | 1 - 2 | 126 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |