Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144868.3 | NP_659117.2 | 18 - 19 | 3891 | 60 | |
GenBank mRNA | AF237953.1 | - | 16 - 17 | 1908 | 60 | |
BC025838.1 | - | 12 - 13 | 1394 | 60 | ||
BC038056.1 | - | 13 - 14 | 1999 | 60 | ||
BC054751.1 | - | 18 - 19 | 3170 | 60 | ||
BC055289.1 | - | 19 - 20 | 3694 | 60 | ||
BC096538.1 | - | 18 - 19 | 3891 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |