Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008230.6 | NP_032256.3 | 2 - 3 | 330 | 113 | |
GenBank mRNA | AF109137.1 | - | 2 - 3 | 240 | 113 | |
AK076384.1 | - | 2 - 3 | 314 | 113 | ||
AK088545.1 | - | 2 - 3 | 317 | 113 | ||
AK133455.1 | - | 2 - 3 | 331 | 113 | ||
AK150168.1 | - | 2 - 3 | 318 | 113 | ||
AK153104.1 | - | 2 - 3 | 318 | 113 | ||
BC052833.1 | - | 2 - 3 | 315 | 30044098 | 113 | |
X57437.1 | - | 2 - 3 | 295 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |