Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clasrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016680.5 | NP_057889.3 | 3 - 4 | 292 | 69 | |
NR_125832.1 | 3 - 4 | 292 | 69 | |||
NR_125833.1 | 3 - 4 | 292 | 69 | |||
NR_125834.1 | 3 - 4 | 292 | 69 | |||
NR_125835.1 | 3 - 4 | 254 | 69 | |||
GenBank mRNA | AB080582.1 | - | 2 - 3 | 155 | 69 | |
AB080583.1 | - | 2 - 3 | 155 | 69 | ||
AF042799.1 | - | 3 - 4 | 257 | 69 | ||
AK009334.1 | - | 3 - 4 | 243 | 69 | ||
AK159443.1 | - | 3 - 4 | 261 | 69 | ||
BC010728.1 | - | 3 - 4 | 243 | 69 | ||
BC051916.1 | - | 3 - 4 | 267 | 5707517 | 69 | |
BC060092.1 | - | 3 - 4 | 265 | 69 | ||
BC079543.1 | - | 3 - 4 | 255 | 69 | ||
BC106129.1 | - | 3 - 4 | 271 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |