Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024222.2 | NP_077184.2 | 11 - 12 | 2077 | 79 | |
GenBank mRNA | AK012153.1 | - | 11 - 12 | 1747 | 79 | |
AK018758.2 | - | 11 - 12 | 1787 | 79 | ||
AK145674.1 | - | 11 - 12 | 1718 | 79 | ||
AK148065.1 | - | 11 - 12 | 1776 | 79 | ||
AK152899.1 | - | 11 - 12 | 2125 | 79 | ||
AK154979.1 | - | 11 - 12 | 1766 | 79 | ||
AK170079.1 | - | 11 - 12 | 1789 | 79 | ||
AX799084.1 | - | 11 - 12 | 1788 | 79 | ||
BC003206.1 | - | 6 - 7 | 789 | 79 | ||
BC013054.1 | - | 1 - 2 | 195 | 79 | ||
BC052433.1 | - | 11 - 12 | 2077 | 6837097 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |