Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbfox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110827.2 | NP_001104297.1 | 12 - 13 | 1737 | 67 | |
NM_001110828.2 | NP_001104298.1 | 11 - 12 | 1291 | 67 | ||
NM_001110829.2 | NP_001104299.1 | 11 - 12 | 1282 | 67 | ||
NM_001110830.2 | NP_001104300.1 | 11 - 12 | 1279 | 67 | ||
NM_001286417.1 | NP_001273346.1 | 10 - 11 | 1239 | 67 | ||
NM_001286418.1 | NP_001273347.1 | 8 - 9 | 997 | 67 | ||
NM_001286419.1 | NP_001273348.1 | 10 - 11 | 1186 | 67 | ||
NM_053104.6 | NP_444334.3 | 12 - 13 | 1749 | 67 | ||
NM_175387.3 | NP_780596.1 | 11 - 12 | 1697 | 67 | ||
GenBank mRNA | AF229055.1 | - | 11 - 12 | 1222 | 67 | |
AF387322.1 | - | 11 - 12 | 1023 | 67 | ||
AK044929.1 | - | 10 - 11 | 1214 | 67 | ||
AK077594.1 | - | 12 - 13 | 1625 | 67 | ||
AK139555.1 | - | 13 - 14 | 1721 | 67 | ||
AY659951.1 | - | 11 - 12 | 1023 | 67 | ||
AY659952.1 | - | 11 - 12 | 1026 | 67 | ||
AY659953.1 | - | 8 - 9 | 741 | 67 | ||
AY904025.1 | - | 13 - 14 | 1173 | 67 | ||
BC002124.1 | - | 11 - 12 | 1117 | 3494630 | 67 | |
BC027263.1 | - | 11 - 12 | 1249 | 3661748 | 67 | |
DQ017388.1 | - | 11 - 12 | 1035 | 67 | ||
DQ017389.1 | - | 10 - 11 | 930 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |