Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145139.2 | NP_660121.2 | 4 - 5 | 414 | 85 | |
GenBank mRNA | AJ310346.1 | - | 4 - 5 | 424 | 85 | |
AK032936.1 | - | 4 - 5 | 402 | 85 | ||
AK075584.1 | - | 4 - 5 | 420 | 85 | ||
AK150895.1 | - | 4 - 5 | 409 | 85 | ||
AK151175.1 | - | 4 - 5 | 409 | 85 | ||
AK167640.1 | - | 4 - 5 | 409 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |