Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153056.2 | NP_694696.2 | 3 - 4 | 368 | 67 | |
GenBank mRNA | AK135484.1 | - | 3 - 4 | 365 | 67 | |
AY251540.1 | - | 3 - 4 | 344 | 67 | ||
BC026403.1 | - | 3 - 4 | 333 | 4209498 | 67 | |
BC026637.1 | - | 3 - 4 | 336 | 67 | ||
BC026650.1 | - | 3 - 4 | 336 | 4987746 | 67 | |
BC040759.1 | - | 3 - 4 | 333 | 4209498 | 67 | |
BC080304.1 | - | 2 - 3 | 672 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |