Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024227.3 | NP_077189.2 | 2 - 3 | 506 | 81 | |
GenBank mRNA | AK003727.2 | - | 2 - 3 | 487 | 81 | |
AK031701.1 | - | 2 - 3 | 476 | 81 | ||
AK046544.1 | - | 2 - 3 | 487 | 81 | ||
AK163190.1 | - | 2 - 3 | 489 | 81 | ||
BC002293.1 | - | 2 - 3 | 477 | 81 | ||
BC005797.1 | - | 2 - 3 | 478 | 81 | ||
BC080753.1 | - | 2 - 3 | 452 | 81 | ||
BC100322.1 | - | 2 - 3 | 478 | 81 | ||
BC145680.1 | - | 2 - 3 | 475 | 40130983 | 81 | |
BC145682.1 | - | 2 - 3 | 475 | 40130985 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |