Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284411.1 | NP_001271340.1 | 1 - 2 | 94 | 63 | |
NM_001284412.1 | NP_001271341.1 | 1 - 2 | 94 | 63 | ||
NM_001284414.1 | NP_001271343.1 | 1 - 2 | 94 | 63 | ||
NM_026776.4 | NP_081052.2 | 1 - 2 | 94 | 63 | ||
GenBank mRNA | AK002500.1 | - | 1 - 2 | 56 | 63 | |
AK003715.2 | - | 1 - 2 | 66 | 63 | ||
AK003877.1 | - | 1 - 2 | 52 | 63 | ||
AK082933.1 | - | 1 - 2 | 73 | 63 | ||
AK145811.1 | - | 1 - 2 | 65 | 63 | ||
AK150822.1 | - | 1 - 2 | 76 | 63 | ||
AK162230.1 | - | 1 - 2 | 67 | 63 | ||
AK166008.1 | - | 1 - 2 | 73 | 63 | ||
BC029181.1 | - | 1 - 2 | 58 | 4240500 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |