Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007934.3 | NP_031960.1 | 12 - 13 | 2185 | 88 | |
GenBank mRNA | AK031439.1 | - | 11 - 12 | 1217 | 88 | |
AK128975.1 | - | 12 - 13 | 2142 | 88 | ||
AK146808.1 | - | 12 - 13 | 2157 | 88 | ||
BC127060.1 | - | 12 - 13 | 2049 | 40109863 | 88 | |
DQ000292.1 | - | 12 - 13 | 1925 | 88 | ||
M29961.1 | - | 12 - 13 | 2004 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |