Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113331.2 | NP_001106802.1 | 2 - 3 | 800 | 63 | |
NM_011368.5 | NP_035498.2 | 3 - 4 | 402 | 63 | ||
GenBank mRNA | AK036980.1 | - | 1 - 2 | 2368 | 63 | |
AK049357.1 | - | 3 - 4 | 375 | 63 | ||
AK155158.1 | - | 2 - 3 | 795 | 63 | ||
BC036172.1 | - | 3 - 4 | 353 | 4988907 | 63 | |
BY341731.1 | - | 2 - 3 | 235 | 63 | ||
U15784.1 | - | 3 - 4 | 292 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons |