Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113331.2 | NP_001106802.1 | 3 - 4 | 854 | 96 | |
NM_011368.5 | NP_035498.2 | 4 - 5 | 456 | 96 | ||
GenBank mRNA | AK036980.1 | - | 2 - 3 | 2422 | 96 | |
AK049357.1 | - | 4 - 5 | 429 | 96 | ||
AK155158.1 | - | 3 - 4 | 849 | 96 | ||
BC036172.1 | - | 4 - 5 | 407 | 4988907 | 96 | |
BY341731.1 | - | 3 - 4 | 289 | 96 | ||
U15784.1 | - | 4 - 5 | 346 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |