Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077705.2 | NP_001071173.1 | 9 - 10 | 1274 | 80 | |
NM_013545.3 | NP_038573.2 | 9 - 10 | 1272 | 80 | ||
GenBank mRNA | AK132509.1 | - | 9 - 10 | 1191 | 80 | |
AK150509.1 | - | 9 - 10 | 1179 | 80 | ||
AK151077.1 | - | 9 - 10 | 1193 | 80 | ||
AK151623.1 | - | 9 - 10 | 1196 | 80 | ||
AK159686.1 | - | 9 - 10 | 1231 | 80 | ||
AK159833.1 | - | 9 - 10 | 1224 | 80 | ||
AK167923.1 | - | 9 - 10 | 1226 | 80 | ||
BC012660.1 | - | 9 - 10 | 1217 | 4010462 | 80 | |
M68902.1 | - | 9 - 10 | 1224 | 80 | ||
M90389.1 | - | 9 - 10 | 1174 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |