Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256130.1 | NP_001243059.1 | 5 - 6 | 791 | 113 | |
NM_001256131.1 | NP_001243060.1 | 4 - 5 | 670 | 113 | ||
NM_001256132.1 | NP_001243061.1 | 5 - 6 | 595 | 113 | ||
NM_001256133.1 | NP_001243062.1 | 3 - 4 | 245 | 113 | ||
NM_023647.6 | NP_076136.2 | 5 - 6 | 1545 | 113 | ||
GenBank mRNA | AB041581.1 | - | 5 - 6 | 728 | 113 | |
AK008106.1 | - | 5 - 6 | 778 | 113 | ||
AK011248.1 | - | 4 - 5 | 615 | 113 | ||
AK012859.1 | - | 5 - 6 | 729 | 113 | ||
AK028734.1 | - | 5 - 6 | 1503 | 113 | ||
AK041427.1 | - | 5 - 6 | 792 | 113 | ||
AK076051.1 | - | 5 - 6 | 748 | 113 | ||
AK076184.1 | - | 5 - 6 | 732 | 113 | ||
AK154755.1 | - | 5 - 6 | 749 | 113 | ||
BC038499.1 | - | 5 - 6 | 733 | 4482150 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |