Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spock3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252620.1 | NP_001239549.1 | 1 - 2 | 76 | 67 | |
NM_001252621.1 | NP_001239550.1 | 1 - 2 | 76 | 67 | ||
GenBank mRNA | AJ278998.1 | - | 1 - 2 | 84 | 67 | |
AK049843.1 | - | 1 - 2 | 77 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |